missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoroacetoacetate, 97%

Catalog No. AAA1371730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1371730 250 g
AAA1371709 10 g
AAA1371718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1371730 Supplier Thermo Scientific Chemicals Supplier No. A1371730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-31-6 | C6H7F3O3 | 184.114 g/mol

Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

Solubility
Miscible with water, ethanol, benzene and organic solvents.

Notes
Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 372-31-6
Molecular Formula C6H7F3O3
Molecular Weight (g/mol) 184.114
MDL Number MFCD00000424
InChI Key OCJKUQIPRNZDTK-UHFFFAOYSA-N
Synonym ethyl 4,4,4-trifluoroacetoacetate, ethyl trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-3-oxo-, ethyl ester, 4,4,4-trifluoro-3-oxobutanoic acid ethyl ester, ethyl trifluoroacetyl acetate, etfaa, 4,4,4-trifluoroacetoacetic acid ethyl ester, ethyl-4,4,4-trifluoroacetoacetate, trifluoroacetoacetic acid ethyl ester, 4,4,4-trifluoro-3-oxo-butyric acid ethyl ester
PubChem CID 67793
IUPAC Name ethyl 4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)C(F)(F)F

Specifications

Melting Point -30°C
Density 1.259
Boiling Point 129°C to 130°C
Flash Point 38°C (100°F)
Odor Odorless
Refractive Index 1.376
Quantity 250 g
UN Number UN3272
Beilstein 608353
Solubility Information Miscible with water,ethanol,benzene and organic solvents.
Formula Weight 184.12
Percent Purity ≥98%
Chemical Name or Material Ethyl trifluoroacetoacetate
Show More Show Less
Hazard Category H226-H302
Hazard Statement GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 206-750-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only