missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoroacetate 99.0+%, TCI America™

Catalog No. p-7136279
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T043225G 25 g
T0432100G 100 g
T0432500G 500 g
3 options

Catalog No. T043225G

Supplier: TCI America T043225G

Only null left

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00000419
InChI Key STSCVKRWJPWALQ-UHFFFAOYSA-N
Synonym ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid
PubChem CID 9794
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 g
UN Number 2924
Formula Weight 142.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trifluoroacetate
EINECSNumber (2)-3532
TSCA Yes