missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoroacetate 99.0+%, TCI America™

Catalog No. T043225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T043225G 25 g
T0432100G 100 g
T0432500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T043225G Supplier TCI America Supplier No. T043225G

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00000419
InChI Key STSCVKRWJPWALQ-UHFFFAOYSA-N
Synonym ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid
PubChem CID 9794
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 g
UN Number 2924
Formula Weight 142.08
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trifluoroacetate
EINECSNumber (2)-3532
TSCA Yes