missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoroacetate, 99%

Catalog No. p-3736507
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119020250 25 mL Glass bottle
AC119021000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119020250 Supplier Thermo Scientific Chemicals Supplier No. 119020250
Only null left

CAS: 383-63-1 | C4H5F3O2 | 142.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.08
MDL Number MFCD00000419
InChI Key STSCVKRWJPWALQ-UHFFFAOYSA-N
Synonym ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid
PubChem CID 9794
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Melting Point -78.0°C
Density 1.1940g/mL
Boiling Point 60.0°C to 62.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3060 to 1.3080
Linear Formula CF3CO2C2H5
Quantity 25 mL
Beilstein 02,II,186
Specific Gravity 1.194
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Formula Weight 142.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl trifluoroacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irrit
EINECSNumber 206-851-6

RUO – Research Use Only