missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trifluoroacetate, 99%

Catalog No. aaa115200b
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1000 g
250 g
50 g
Catalog No. Quantity
AAA115200B 1000 g
AAA1152018 50 g
AAA1152030 250 g
3 options

Catalog No. AAA115200B

Supplier: Thermo Scientific Chemicals A115200B

Only null left

CAS: 383-63-1 | C4H5F3O2 | 142.077 g/mol

Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.

Solubility
Slightly miscible with water. Miscible with chloroform and methanol.

Notes
Moisture Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.

Chemical Identifiers

CAS 383-63-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.077
MDL Number MFCD00000419
InChI Key STSCVKRWJPWALQ-UHFFFAOYSA-N
Synonym ethyl trifluoroacetate, trifluoroacetic acid ethyl ester, ethyltrifluoroacetate, acetic acid, trifluoro-, ethyl ester, trifluoroacetic acid, ethyl ester, ethyl trifluoroethanoate, unii-a6tzk6x11x, acetic acid, 2,2,2-trifluoro-, ethyl ester, a6tzk6x11x, ethyl ester of trifluoroacetic acid
PubChem CID 9794
IUPAC Name ethyl 2,2,2-trifluoroacetate
SMILES CCOC(=O)C(F)(F)F

Specifications

Melting Point <-78°C
Density 1.192
Boiling Point 60°C to 62°C
Flash Point −1°C (30°F)
Odor Sweet
Refractive Index 1.307
Linear Formula CF3CO2CH2CH3
Quantity 1000 g
UN Number UN2924
Beilstein 1761411
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water. Miscible with chloroform and methanol.
Formula Weight 142.08
Percent Purity 99%
Chemical Name or Material Ethyl trifluoroacetate
Show More Show Less
Hazard Category H225-H319
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-851-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only