missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trichloroacetate 97.0+%, TCI America™

Catalog No. T150425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T150425G 25 g
T1504500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T150425G Supplier TCI America Supplier No. T150425G
Only null left

Chemical Identifiers

CAS 515-84-4
Molecular Formula C4H5Cl3O2
Molecular Weight (g/mol) 191.43
MDL Number MFCD00000795
InChI Key SJMLNDPIJZBEKY-UHFFFAOYSA-N
Synonym ethyl trichloroacetate, acetic acid, trichloro-, ethyl ester, unii-br8fkn5z55, acetic acid, ester with trichloroethanol, ethyltrichloroacetate, br8fkn5z55, acetic acid, 2,2,2-trichloro-, ethyl ester, acmc-209kup, ethyl-2,2,2-trichloroacetate
PubChem CID 10588
IUPAC Name ethyl 2,2,2-trichloroacetate
SMILES CCOC(=O)C(Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 167°C
Quantity 25 g
Formula Weight 191.43
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trichloroacetate
EINECSNumber (2)-3606
TSCA Yes