missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-Hydroxycyclohexanecarboxylate 98.0+%, TCI America™

$432.46 - $1081.16

Chemical Identifiers

CAS 3618-04-0
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
InChI Key BZKQJSLASWRDNE-UHFFFAOYSA-N
Synonym trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester
PubChem CID 86973
IUPAC Name ethyl 4-hydroxycyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCC(CC1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E08385G
View Documents
TCI America
E08385G
5 g
Each for $432.46
Only null left
 
E083825G
View Documents
TCI America
E083825G
25 g
Each for $1,081.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3618-04-0
172.224
trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester
ethyl 4-hydroxycyclohexane-1-carboxylate
C9H16O3
BZKQJSLASWRDNE-UHFFFAOYSA-N
86973
CCOC(=O)C1CCC(CC1)O

Specifications

3618-04-0
144°C
5 g
BZKQJSLASWRDNE-UHFFFAOYSA-N
ethyl 4-hydroxycyclohexane-1-carboxylate
86973
≥98.0% (GC)
Ethyl trans-4-Hydroxycyclohexanecarboxylate
Yellow
C9H16O3
trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester
CCOC(=O)C1CCC(CC1)O
172.224
172.22
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'