missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-carboxylate, 98%

Catalog No. AAB2246803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2246803 1 g
AAB2246806 5 g
AAB2246814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2246803 Supplier Thermo Scientific Chemicals Supplier No. B2246803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5751-80-4 | C7H8O2S | 156.199 g/mol

Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 5751-80-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD02179226
InChI Key OYSLMAQEMAJMCL-UHFFFAOYSA-N
Synonym ethyl 3-thenoate, ethyl 3-thiophenecarboxylate, ethylthiophene-3-carboxylate, 3-ethoxycarbonyl thiophene, 3-thiophenecarboxylic acid ethyl ester, 3-thiophenecarboxylic acid, ethyl ester, thiophene-3-carboxylic acid ethyl ester, pubchem16116, ksc493o3n
PubChem CID 79824
IUPAC Name ethyl thiophene-3-carboxylate
SMILES CCOC(=O)C1=CSC=C1

Specifications

Boiling Point 207°C to 208°C
Refractive Index 1.5225
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 156.2
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-carboxylate
EINECSNumber 227-275-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only