missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-acetate, 98%

Catalog No. AAA1969422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1969422 100 g
AAA1969406 5 g
AAA1969414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1969422 Supplier Thermo Scientific Chemicals Supplier No. A1969422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37784-63-7 | C8H10O2S | 170.23 g/mol

Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 37784-63-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00005472
InChI Key FZBNQIWPYCUPAP-UHFFFAOYSA-N
Synonym ethyl thiophene-3-acetate, ethyl 3-thienylacetate, ethyl 3-thiopheneacetate, ethyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, ethyl ester, ethyl 3-thienylacetate #, acmc-1afw1, ethyl 2-3-thienyl acetate, bidd:gt0101, ethyl 2-thiophen-3-ylethanoate
PubChem CID 520865
SMILES CCOC(=O)CC1=CSC=C1

Specifications

Density 1.137
Boiling Point 110°C to 114°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.51
Quantity 100 g
Beilstein 122555
IUPAC Name ethyl 2-thiophen-3-ylacetate
Formula Weight 170.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl thiophene-3-acetate
Show More Show Less
EINECSNumber 253-665-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only