missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-3-acetate, 98%

Catalog No. AAA1969406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1969406 5 g
AAA1969414 25 g
AAA1969422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1969406 Supplier Thermo Scientific Chemicals Supplier No. A1969406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37784-63-7 | C8H10O2S | 170.23 g/mol

Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-acetate is used in the syntheses of poly(thiophene-3-acetic acid) and solution-processable polymers, which consist of polythiophene main chain and alkyl and oligoaniline side groups.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 37784-63-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00005472
InChI Key FZBNQIWPYCUPAP-UHFFFAOYSA-N
Synonym ethyl thiophene-3-acetate, ethyl 3-thienylacetate, ethyl 3-thiopheneacetate, ethyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, ethyl ester, ethyl 3-thienylacetate #, acmc-1afw1, ethyl 2-3-thienyl acetate, bidd:gt0101, ethyl 2-thiophen-3-ylethanoate
PubChem CID 520865
SMILES CCOC(=O)CC1=CSC=C1

Specifications

Density 1.137
Boiling Point 110°C to 114°C (6 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.51
Quantity 5 g
Beilstein 122555
IUPAC Name ethyl 2-(thiophen-3-yl)acetate
Formula Weight 170.23
Percent Purity 98%
Chemical Name or Material Ethyl thiophene-3-acetate
EINECSNumber 253-665-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only