missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiooxamate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 319600010

 View more versions of this product

Catalog No. AC319600010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl thiooxamate
Authentic
Glass bottle
H2NCSCO2C2H5
1g
YMBMCMOZIGSBOA-UHFFFAOYSA-N
ethyl 2-amino-2-sulfanylideneacetate
2733398
95%
16982-21-1
94% min. (GC)
C4H7NO2S
MFCD00074903
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
CCOC(=O)C(=S)N
133.17
133.17

Chemical Identifiers

16982-21-1
133.17
YMBMCMOZIGSBOA-UHFFFAOYSA-N
2733398
CCOC(=O)C(=S)N
C4H7NO2S
MFCD00074903
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
ethyl 2-amino-2-sulfanylideneacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only