missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Thioglycolate 97.0+%, TCI America™

Catalog No. T021125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T021125G 25 g
T0211500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T021125G Supplier TCI America Supplier No. T021125G

Chemical Identifiers

CAS 623-51-8
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00004874
InChI Key PVBRSNZAOAJRKO-UHFFFAOYSA-N
Synonym ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester
PubChem CID 12185
IUPAC Name ethyl 2-sulfanylacetate
SMILES CCOC(=O)CS

Specifications

Melting Point -80°C
Color Yellow
Boiling Point 157°C
Quantity 25 g
UN Number 1992
Formula Weight 120.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Thioglycolate
EINECSNumber (2)-1358
RTECSNumber AI6650000
TSCA Yes