missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tert-Butylacetate 99.0+%, TCI America™

Catalog No. B151025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B151025ML 25 mL
B1510100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B151025ML Supplier TCI America Supplier No. B151025ML

Chemical Identifiers

CAS 5340-78-3
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00026911
InChI Key JWMNHAMYTBAUPI-UHFFFAOYSA-N
Synonym ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
PubChem CID 79284
IUPAC Name ethyl 3,3-dimethylbutanoate
SMILES CCOC(=O)CC(C)(C)C

Specifications

Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 3272
Formula Weight 144.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl tert-Butylacetate
EINECSNumber (2)-0782
TSCA No