Learn More
Ethyl tert-butylacetate, 98%
CAS: 5340-78-3 | C8H16O2 | 144.214 g/mol
Supplier: Thermo Scientific Chemicals L0560314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl tert-butylacetate | |
| 0.86 | |
| 34°C (93°F) | |
| 1.4035 | |
| 25 g | |
| 1749778 | |
| JWMNHAMYTBAUPI-UHFFFAOYSA-N | |
| ethyl 3,3-dimethylbutanoate | |
| 79284 | |
| 98% |
| 5340-78-3 | |
| 144°C to 146°C | |
| C8H16O2 | |
| MFCD00026911 | |
| UN3272 | |
| ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester | |
| CCOC(=O)CC(C)(C)C | |
| 144.214 | |
| 144.22 |
Chemical Identifiers
| 5340-78-3 | |
| 144.214 | |
| JWMNHAMYTBAUPI-UHFFFAOYSA-N | |
| 79284 | |
| CCOC(=O)CC(C)(C)C |
| C8H16O2 | |
| MFCD00026911 | |
| ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester | |
| ethyl 3,3-dimethylbutanoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 226-280-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only