missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tert-butylacetate, 98%

Catalog No. AAL0560306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0560306 5 g
AAL0560314 25 g
AAL0560322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0560306 Supplier Thermo Scientific Chemicals Supplier No. L0560306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5340-78-3 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5340-78-3
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00026911
InChI Key JWMNHAMYTBAUPI-UHFFFAOYSA-N
Synonym ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
PubChem CID 79284
IUPAC Name ethyl 3,3-dimethylbutanoate
SMILES CCOC(=O)CC(C)(C)C

Specifications

Density 0.86
Boiling Point 144°C to 146°C
Flash Point 34°C (93°F)
Refractive Index 1.4035
Quantity 5 g
UN Number UN3272
Beilstein 1749778
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material Ethyl tert-butylacetate
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 226-280-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only