missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Succinyl Chloride 95.0+%, TCI America™

Catalog No. E036825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E036825G 25 g
E0368500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E036825G Supplier TCI America Supplier No. E036825G

Chemical Identifiers

CAS 14794-31-1
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00000751
InChI Key IXZFDJXHLQQSGQ-UHFFFAOYSA-N
Synonym ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride
PubChem CID 84648
IUPAC Name ethyl 4-chloro-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)Cl

Specifications

Color Yellow
Boiling Point 144°C
Quantity 25 g
UN Number 3265
Formula Weight 164.59
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl Succinyl Chloride