missing translation for 'onlineSavingsMsg'
Learn More

Ethyl succinyl chloride, 95%, Thermo Scientific™

Catalog No. p-132603
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC184090250 25g Glass bottle
AC184091000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC184090250 Supplier Thermo Scientific Chemicals Supplier No. 184090250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 14794-31-1
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000751
InChI Key IXZFDJXHLQQSGQ-UHFFFAOYSA-N
Synonym ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride
PubChem CID 84648
IUPAC Name ethyl 4-chloro-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)Cl

Specifications

Density 1.1575g/mL
Boiling Point 88.0°C to 90.0°C (11.0mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4390
Linear Formula ClCOCH2CH2CO2C2H5
Quantity 25g
Beilstein 02, 613
Specific Gravity 1.1575
Formula Weight 164.59
Percent Purity 95%
Chemical Name or Material Ethyl succinyl chloride, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 238-855-9
TSCA TSCA

RUO – Research Use Only