missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Stilbene-4-carboxylate 98.0+%, TCI America™

Catalog No. p-7129102
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06781G 1 g
E06785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06781G Supplier TCI America Supplier No. E06781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109463-48-1
Molecular Formula C17H16O2
Molecular Weight (g/mol) 252.313
MDL Number MFCD06204340
InChI Key RBQGAPDZGPQIDL-CMDGGOBGSA-N
Synonym Stilbene-4-carboxylic Acid Ethyl Ester, 4-(Ethoxycarbonyl)stilbene
PubChem CID 5907907
IUPAC Name ethyl 4-[(E)-2-phenylethenyl]benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C=CC2=CC=CC=C2

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 252.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Stilbene-4-carboxylate