missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Sorbate 98.0+%, TCI America™

Catalog No. p-7135785
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
S005525ML 25 mL
S0055100ML 100 mL
S0055500ML 500 mL
3 options

Catalog No. S005525ML

Supplier: TCI America S005525ML

Only null left

Chemical Identifiers

CAS 2396-84-1
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00009296
InChI Key OZZYKXXGCOLLLO-TWTPFVCWSA-N
Synonym ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459
PubChem CID 1550470
IUPAC Name ethyl (2E,4E)-hexa-2,4-dienoate
SMILES CCOC(=O)\C=C\C=C\C

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 mL
Formula Weight 140.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Sorbate
EINECSNumber (2)-1081
RTECSNumber WG2143000
TSCA Yes