missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Sorbate 98.0+%, TCI America™

Catalog No. S005525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005525ML 25 mL
S0055100ML 100 mL
S0055500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S005525ML Supplier TCI America Supplier No. S005525ML

Chemical Identifiers

CAS 2396-84-1
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00009296
InChI Key OZZYKXXGCOLLLO-TWTPFVCWSA-N
Synonym ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459
PubChem CID 1550470
IUPAC Name ethyl (2E,4E)-hexa-2,4-dienoate
SMILES CCOC(=O)\C=C\C=C\C

Specifications

Color Yellow
Boiling Point 76°C
Quantity 25 mL
Formula Weight 140.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Sorbate
EINECSNumber (2)-1081
RTECSNumber WG2143000
TSCA Yes