missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-3-Piperidinecarboxylate 98.0+%, TCI America™

Catalog No. p-7133931
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
N06795G 5 g
N067925G 25 g
2 options

Catalog No. N06795G

Supplier: TCI America N06795G

Only null left

Chemical Identifiers

CAS 37675-18-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00792499
InChI Key XIWBSOUNZWSFKU-ZETCQYMHSA-N
Synonym s-ethyl piperidine-3-carboxylate, ethyl 3s-piperidine-3-carboxylate, ethyl s-nipecotate, s-+-nipecotic acid ethyl ester, s-ethyl nipecotate, ethyl s-piperidine-3-carboxylate, 3-piperidinecarboxylic acid, ethyl ester, 3s, s-+-ethyl nipecotate, ethyl s-+-nipecotate, s-nipecotic acid ethyl ester
PubChem CID 187784
IUPAC Name ethyl (3S)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(+)-3-Piperidinecarboxylate