missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-3-Hydroxybutyrate 96.0+%, TCI America™

Catalog No. h09755g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09755G 5 g
H097525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09755G Supplier TCI America Supplier No. H09755G
Only null left

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(+)-3-Hydroxybutyrate
EINECSNumber (2)-1430
TSCA No