missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (R)-nipecotate L-tartrate, 98%, Thermo Scientific Chemicals
$550.78 - $550.78
Chemical Identifiers
| CAS | 167392-57-6 |
|---|---|
| Molecular Formula | C12H21NO8 |
| Molecular Weight (g/mol) | 307.299 |
| MDL Number | MFCD00799538 |
| InChI Key | HHPGQKZOPPDLNH-RATVZYDNSA-N |
| Synonym | s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester |
| PubChem CID | 53249980 |
| IUPAC Name | (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate |
| SMILES | CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5622614
|
Thermo Scientific Chemicals
H5622614 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 167392-57-6 | |
| 307.299 | |
| HHPGQKZOPPDLNH-RATVZYDNSA-N | |
| 53249980 | |
| CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O |
| C12H21NO8 | |
| MFCD00799538 | |
| s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester | |
| (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate |
Specifications
| 167392-57-6 | |
| C12H21NO8 | |
| 25 g | |
| HHPGQKZOPPDLNH-RATVZYDNSA-N | |
| CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O | |
| 307.299 | |
| 307.3 | |
| Ethyl (R)-nipecotate L-tartrate |
| 157°C to 159°C | |
| MFCD00799538 | |
| s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester | |
| +10° (c=5 in Water) | |
| (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate | |
| 53249980 | |
| 98% |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only