missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-nipecotate L-tartrate, 98%, Thermo Scientific Chemicals

Catalog No. AAH5622614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5622614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5622614 Supplier Thermo Scientific Chemicals Supplier No. H5622614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 167392-57-6 | Molecular Formula: C12H21NO8 | Molecular Weight: 307.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167392-57-6
Molecular Formula C12H21NO8
Molecular Weight (g/mol) 307.299
MDL Number MFCD00799538
InChI Key HHPGQKZOPPDLNH-RATVZYDNSA-N
Synonym s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
PubChem CID 53249980
IUPAC Name (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 157°C to 159°C
Quantity 25 g
Optical Rotation +10° (c=5 in Water)
Formula Weight 307.3
Percent Purity 98%
Chemical Name or Material Ethyl (R)-nipecotate L-tartrate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only