missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(-)-4-Cyano-3-hydroxybutyrate 97.0+%, TCI America™

Catalog No. p-7125003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14741G 1 g
C14745G 5 g
C147425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C14741G Supplier TCI America Supplier No. C14741G
Only null left

Chemical Identifiers

CAS 141942-85-0
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00270839
InChI Key LOQFROBMBSKWQY-ZCFIWIBFSA-N
Synonym ethyl r---4-cyano-3-hydroxybutyate, ethyl r---4-cyano-3-hydroxybutyrate, ethyl 3r-4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxybutyric acid ethyl ester, r-ethyl 4-cyano-3-hydroxybutyrate, ethyl-r-+-4-cyano-3-hydroxybutanoate, ethyl r-4-cyano-3-hydroxybutyrate, r-ethyl 4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxy-butyric acid ethyl ester, r---4-cyano-3-hydroxybutyric acid ethyl ester
PubChem CID 2733879
IUPAC Name ethyl (3R)-4-cyano-3-hydroxybutanoate
SMILES CCOC(=O)C[C@H](O)CC#N

Specifications

Color Yellow
Boiling Point 270°C
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (R)-(-)-4-Cyano-3-hydroxybutyrate