missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-3-Piperidinecarboxylate L-Tartrate 98.0+%, TCI America™

Catalog No. p-7133932
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
N06805G 5 g
N068025G 25 g
2 options

Catalog No. N06805G

Supplier: TCI America N06805G

Only null left

Chemical Identifiers

CAS 167392-57-6
Molecular Formula C12H21NO8
Molecular Weight (g/mol) 307.299
MDL Number MFCD00799538
InChI Key HHPGQKZOPPDLNH-RATVZYDNSA-N
Synonym s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate, 3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
PubChem CID 53249980
IUPAC Name (3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 157°C
Color White
Quantity 5 g
Formula Weight 307.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl (R)-3-Piperidinecarboxylate L-Tartrate