missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific™

Catalog No. p-7038120
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
AAH5701903 1 g
1 options

Catalog No. AAH5701903

Supplier: Thermo Scientific Chemicals H5701903

Only null left

It is used as pharmaceutical intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.

Chemical Identifiers

CAS 357154-16-6
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD20526405
InChI Key YAVQLRUBKDCOCI-SNVBAGLBSA-N
Synonym r-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, ethyl r-1-boc-4-oxopiperidine-2-carboxylate, r-+-1-boc-4-oxopiperidine-2-carboxylic acid ethyl ester, r-1-boc-4-oxopiperidine-2-carboxylic acid ethyl ester, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl r-+-1-boc-4-oxopipecolate, r-1-tert-butyl-2-ethyl-4-oxopiperidine-1,2-dicarboxylate, 2r-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester, 1-tert-butyl 2-ethyl 2r-4-oxopiperidine-1,2-dicarboxylate
PubChem CID 67586303
IUPAC Name 1-O-tert-butyl 2-O-ethyl (2R)-4-oxopiperidine-1,2-dicarboxylate
SMILES CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Solubility Information Soluble in chloroform and methanol.
Optical Rotation +20° (c=0.5 in Chloroform)
Formula Weight 271.31
Percent Purity 95%
Chemical Name or Material Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only