missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pyrimidine-4-carboxylate 98.0+%, TCI America™

Catalog No. E08815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08815G 5 g
E088125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08815G Supplier TCI America Supplier No. E08815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62846-82-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD09832999
InChI Key DWRWSNAREGLUHZ-UHFFFAOYSA-N
Synonym ethyl 4-pyrimidinecarboxylate, ethyl4-pyrimidinecarboxylate, pyrimidine-4-carboxylic acid ethyl ester, 4-pyrimidinecarboxylic acid, ethyl ester, pubchem20807, 4-ethoxycarbonylpyrimidine, ethylpyrimidine-4-carboxylate, 2-ethylpyrimidine-4-carboxylate, 4-pyrimidinecarboxylicacid, ethyl ester, pyrimidine-6-carboxylic acid ethyl ester
PubChem CID 13639438
IUPAC Name ethyl pyrimidine-4-carboxylate
SMILES CCOC(=O)C1=NC=NC=C1

Specifications

Melting Point 39°C
Color White-Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl Pyrimidine-4-carboxylate