missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Pyrazole-4-carboxylate 98.0+%, TCI America™
$76.43 - $218.10
Chemical Identifiers
| CAS | 37622-90-5 |
|---|---|
| Molecular Formula | C6H8N2O2 |
| Molecular Weight (g/mol) | 140.142 |
| MDL Number | MFCD00010844 |
| InChI Key | KACZQOKEFKFNDB-UHFFFAOYSA-N |
| PubChem CID | 142179 |
| IUPAC Name | ethyl 1H-pyrazole-4-carboxylate |
| SMILES | CCOC(=O)C1=CNN=C1 |
Chemical Identifiers
| 37622-90-5 | |
| 140.142 | |
| KACZQOKEFKFNDB-UHFFFAOYSA-N | |
| ethyl 1H-pyrazole-4-carboxylate |
| C6H8N2O2 | |
| MFCD00010844 | |
| 142179 | |
| CCOC(=O)C1=CNN=C1 |
Specifications
| 37622-90-5 | |
| White | |
| C6H8N2O2 | |
| 1 g | |
| KACZQOKEFKFNDB-UHFFFAOYSA-N | |
| ethyl 1H-pyrazole-4-carboxylate | |
| 142179 | |
| ≥98.0% (GC) | |
| Ethyl 4-Pyrazolecarboxylate |
| 78°C | |
| 140°C | |
| MFCD00010844 | |
| ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug | |
| CCOC(=O)C1=CNN=C1 | |
| 140.142 | |
| 140.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No