missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pyrazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E08715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08715G 5 g
E08711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08715G Supplier TCI America Supplier No. E08715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37622-90-5
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00010844
InChI Key KACZQOKEFKFNDB-UHFFFAOYSA-N
PubChem CID 142179
IUPAC Name ethyl 1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=CNN=C1

Specifications

Melting Point 78°C
Color White
Boiling Point 140°C
Quantity 5 g
Synonym ethyl 4-pyrazolecarboxylate, ethyl pyrazole-4-carboxylate, 1h-pyrazole-4-carboxylic acid ethyl ester, 4-ethoxycarbonylpyrazole, 1h-pyrazole-4-carboxylic acid, ethyl ester, 4-pyrazolecarboxylic acid ethyl ester, ethyl4-pyrazolecarboxylate, zlchem 132, pubchem21939, acmc-209iug
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Pyrazole-4-carboxylate