missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propionate 99.0+%, TCI America™

Catalog No. p-7134645
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
P050525ML 25 mL
P0505500ML 500 mL
2 options

Catalog No. P050525ML

Supplier: TCI America P050525ML

Only null left

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Color Colorless
Boiling Point 99°C
Quantity 25 mL
UN Number 1195
Formula Weight 102.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Propionate
EINECSNumber (2)-0774
RTECSNumber UF3675000
TSCA Yes