missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionate, 99+%

Catalog No. AC150505000
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150505000 500 mL Glass bottle
AC150500250 25 mL Glass bottle
AC150500010 1 L Glass bottle
AC150500025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150505000 Supplier Thermo Scientific Chemicals Supplier No. 150505000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-37-3 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point -73°C
Color Colorless to Yellow
Density 0.8870g/mL
Boiling Point 99°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3830 to 1.387
Linear Formula C2H5CO2C2H5
Quantity 500 mL
Beilstein 02, 240
Fieser 01,385
Specific Gravity 0.887
Merck Index 15, 3899
Solubility Information Solubility in water: 25g/L water (15°C). Other solubilities: miscible with alcohol,ether and propylene glycol
Viscosity <1 mPa.s (20°C)
Formula Weight 102.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl propionate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-291-4

RUO – Research Use Only