missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionate, 99%

Catalog No. aaa16650ak
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16650AK 250 mL
AAA16650AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16650AK Supplier Thermo Scientific Chemicals Supplier No. A16650AK
Only null left

CAS: 105-37-3 | C5H10O2 | 102.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00009308
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Melting Point -74°C
Density 0.891
Boiling Point 99°C
Flash Point 12°C (54°F)
Refractive Index 1.385
Quantity 250 mL
UN Number UN1195
Beilstein 506287
Merck Index 14,3847
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material Ethyl propionate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL PROPIONATE
EINECSNumber 203-291-4
RTECSNumber UF3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only