missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propenyl ether, cis + trans, 97%

CAS: 928-55-2 | C5H10O | 86.13 g/mol

Supplier:  Thermo Scientific Chemicals H2631914

 View more versions of this product

Catalog No. AAH2631914


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl propenyl ether
928-55-2
69°C to 71°C
C5H10O
MFCD00015184
UN3271
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
CCO\C=C/C
86.13
86.13
cis + trans
0.778
20°C (68°F)
1.398
25 g
1735974
XDHOEHJVXXTEDV-HYXAFXHYSA-N
(E)-1-ethoxyprop-1-ene
5365091
97%

Chemical Identifiers

928-55-2
86.13
XDHOEHJVXXTEDV-HYXAFXHYSA-N
5365091
C5H10O
MFCD00015184
1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
CCO\C=C/C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 213-176-0

RTECSNumber : KO0525000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only