missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Potassium Oxalate 95.0+%, TCI America™

Catalog No. E11845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11845G 5 g
E118425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11845G Supplier TCI America Supplier No. E11845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1906-57-6
Molecular Formula C4H5KO4
Molecular Weight (g/mol) 156.178
MDL Number MFCD00045861
InChI Key RLPQQBNSTHRHEK-UHFFFAOYSA-M
Synonym ethyl potassium oxalate, potassium ethyl oxalate, ethanedioic acid, monoethyl ester, potassium salt, potassium 2-ethoxy-2-oxoacetate, oxalic acid 1-ethyl 2-potassium salt, ethanedioic acid, 1-ethyl ester, potassium salt 1:1, potassium ethoxy oxo acetate, ambkkkkk567, oxalic acid, monoethyl ester, potassium salt, potassium monoethyl oxalate
PubChem CID 23678856
IUPAC Name potassium;2-ethoxy-2-oxoacetate
SMILES CCOC(=O)C(=O)[O-].[K+]

Specifications

Melting Point 220°C
Color White
Quantity 5 g
Formula Weight 156.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Potassium Oxalate