missing translation for 'onlineSavingsMsg'
Learn More

Ethyl picolinoylacetate, 95%

Catalog No. AC425470025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425470025 2.5 g Glass bottle
AC425470100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425470025 Supplier Thermo Scientific Chemicals Supplier No. 425470025

CAS: 26510-52-1 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26510-52-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094022
InChI Key FQHXWZMJALFSJJ-UHFFFAOYSA-N
Synonym ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
PubChem CID 2736461
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=N1

Specifications

Color Brown to Straw-Yellow
Density 1.1640g/mL
Boiling Point 100°C to 104°C (0.3 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.519
Quantity 2.5 g
Beilstein 22,305
Specific Gravity 1.164
Solubility Information Solubility in water: negligible
Formula Weight 193.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl picolinoylacetate
Show More Show Less

RUO – Research Use Only