missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pentafluoropropionate, 98+%

Catalog No. AAL0178214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0178214 25 g
AAL0178206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0178214 Supplier Thermo Scientific Chemicals Supplier No. L0178214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 426-65-3 | C5H5F5O2 | 192.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 426-65-3
Molecular Formula C5H5F5O2
Molecular Weight (g/mol) 192.085
MDL Number MFCD00000431
InChI Key DBOFMRQAMAZKQY-UHFFFAOYSA-N
Synonym ethyl pentafluoropropionate, ethyl perfluoropropionate, ethylpentafluoropropionate, ethyl pentafluoropropanoate, pentafluoropropionic acid ethyl ester, propanoic acid, pentafluoro-, ethyl ester, propanoic acid, 2,2,3,3,3-pentafluoro-, ethyl ester, 2,2,3,3,3-pentafluoro-propionic acid ethyl ester, pubchem12575, acmc-1asrx
PubChem CID 67928
IUPAC Name ethyl 2,2,3,3,3-pentafluoropropanoate
SMILES CCOC(=O)C(C(F)(F)F)(F)F

Specifications

Density 1.299
Boiling Point 73°C to 74°C
Flash Point 1°C (33°F)
Refractive Index 1.301
Quantity 25 g
UN Number UN3272
Beilstein 1779789
Formula Weight 192.09
Percent Purity ≥98%
Chemical Name or Material Ethyl pentafluoropropionate
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 207-043-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only