missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. T026825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T026825G 25 g
T0268500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T026825G Supplier TCI America Supplier No. T026825G

Chemical Identifiers

CAS 80-40-0
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.252
MDL Number MFCD00009100
InChI Key VRZVPALEJCLXPR-UHFFFAOYSA-N
Synonym ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester
PubChem CID 6638
IUPAC Name ethyl 4-methylbenzenesulfonate
SMILES CCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color White-Yellow
Boiling Point 173°C
Quantity 25 g
Formula Weight 200.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl p-Toluenesulfonate
EINECSNumber (3)-1896
RTECSNumber XT6825000
TSCA Yes