missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxazole-5-carboxylate, 98%

Catalog No. AAH3292406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3292406 5 g
AAH3292414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3292406 Supplier Thermo Scientific Chemicals Supplier No. H3292406

missing translation for 'validMainframeMsg'

CAS: 118994-89-1 | C6H7NO3 | 141.126 g/mol

Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl oxazole-5-carboxylate is used as a pharmaceutical, organic intermediate and in organic light-emitting diode.

Solubility
Soluble in water (35 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 118994-89-1
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD04114930
InChI Key KRMORCCAHXFIHF-UHFFFAOYSA-N
Synonym ethyl oxazole-5-carboxylate, 5-oxazolecarboxylic acid, ethyl ester, 5-oxazolecarboxylicacid, ethyl ester, oxazole-5-carboxylic acid ethyl ester, ethyloxazole-5-carboxylate, 5-ethoxycarbonyloxazole, acmc-1c1iz, ksc510s9l, ethyl 5-oxazolecarboxylate, 5-ethoxycarbonyl-1,3-oxazole
PubChem CID 10964603
IUPAC Name ethyl 1,3-oxazole-5-carboxylate
SMILES CCOC(=O)C1=CN=CO1

Specifications

Boiling Point 60°C (2.25 mmHg)
Refractive Index 1.469
Quantity 5 g
Beilstein 83084416
Solubility Information Soluble in water (35g/L at 25°C).
Formula Weight 141.13
Percent Purity 98%
Chemical Name or Material Ethyl oxazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only