missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Orange, TCI America™

Catalog No. E015525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E015525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E015525G Supplier TCI America Supplier No. E015525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62758-12-7
Molecular Formula C16H18N3NaO3S
Molecular Weight (g/mol) 355.388
MDL Number MFCD00007503
InChI Key YZORUOZKRBVLEG-UHFFFAOYSA-M
Synonym ethyl orange sodium salt, ethyl orange, benzenesulfonic acid, 4-4-diethylamino phenyl azo-, sodium salt, benzenesulfonic acid, 4-2-4-diethylamino phenyl diazenyl-, sodium salt 1:1, sodium p-p-diethylamino phenyl azo benzenesulphonate, 4-4-diethylamino phenyl diazenyl benzenesulfonic acid, sodium salt, 4-diethylaminoazobenzene-4'-sulfonic acid sodium salt, 4'-diethylamino azobenzene-4-sulfonic acid sodium salt
PubChem CID 23676309
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 355.39
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Orange