missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oleate, tech. 70%

Catalog No. p-7022708
Change view
Click to view available options
Quantity:
1000 g
250 g
5000 g
Catalog No. Quantity
AAA1560130 250 g
AAA156010B 1000 g
AAA156010I 5000 g
3 options

Catalog No. AAA1560130

Supplier: Thermo Scientific Chemicals A1560130

Only null left

CAS: 111-62-6 | C20H38O2 | 310.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00009579
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Melting Point -32°C
Density 0.87
Boiling Point 205°C to 208°C
Flash Point >110°C (230°F)
Refractive Index 1.451
Linear Formula CH3(CH2)7CH=CH(CH2)7CO2CH2CH3
Quantity 250 g
Beilstein 1727318
Sensitivity Air Sensitive
Formula Weight 310.53
Percent Purity 70%
Grade Technical
Chemical Name or Material Ethyl oleate
Show More Show Less
EINECSNumber 203-889-5
RTECSNumber RG3715000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only