missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate 95.0+%, TCI America™

Catalog No. O01435ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01435ML 5 mL
O014325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01435ML Supplier TCI America Supplier No. O01435ML

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00009579
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 209°C
Quantity 5 mL
Formula Weight 310.52
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Oleate
EINECSNumber (2)-0798
RTECSNumber RG3715000
TSCA Yes