missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate 70.0+%, TCI America™

Catalog No. p-7134158
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
O005425ML 25 mL
O0054500ML 500 mL
2 options

Catalog No. O005425ML

Supplier: TCI America O005425ML

Only null left

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00009579
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 209°C
Quantity 25 mL
Formula Weight 310.52
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Oleate
EINECSNumber (2)-0798
RTECSNumber RG3715000
TSCA Yes