missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate 70.0+%, TCI America™

Catalog No. O0054500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0054500ML 500 mL
O005425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0054500ML Supplier TCI America Supplier No. O0054500ML

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00009579
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 209°C
Quantity 500 mL
Formula Weight 310.52
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Oleate
EINECSNumber (2)-0798
RTECSNumber RG3715000
TSCA Yes