missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate, NF, Spectrum™ Chemical

Catalog No. p-7157383
Change view
Click to view available options
Quantity:
100 g
25 g
5 kg
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18602543 25 g Amber Glass Bottle
18602544 100 g Amber Glass Bottle
18602545 500 g Amber Glass Bottle
18602546 5 kg Poly Pail
4 options

Catalog No. 18602543

Supplier: Spectrum Chemical Mfg Cor E117225GM

Only null left

E1172, 111-62-6, C20H38O2

Spectrum™ Chemical Ethyl Oleate, NF is used as a solubilizing agent in pharmaceutical preparations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
InChI Key LVGKNOAMLMIIKO-VAWYXSNFSA-N
IUPAC Name ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)OCC

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Grade NF