missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate, NF, Spectrum™ Chemical

Catalog No. 18602543
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602543 25 g Amber Glass Bottle
18602544 100 g Amber Glass Bottle
18602545 500 g Amber Glass Bottle
18602546 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602543 Supplier Spectrum Chemical Mfg Cor Supplier No. E117225GM
Only null left

E1172, 111-62-6, C20H38O2

Spectrum™ Chemical Ethyl Oleate, NF is used as a solubilizing agent in pharmaceutical preparations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
InChI Key LVGKNOAMLMIIKO-VAWYXSNFSA-N
IUPAC Name ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C\CCCCCCCC(=O)OCC

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Grade NF