missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Nonanoate 95.0+%, TCI America™
$48.13 - $215.62
Chemical Identifiers
| CAS | 123-29-5 |
|---|---|
| Molecular Formula | C11H22O2 |
| Molecular Weight (g/mol) | 186.30 |
| MDL Number | MFCD00009570 |
| InChI Key | BYEVBITUADOIGY-UHFFFAOYSA-N |
| Synonym | ethyl pelargonate, nonanoic acid, ethyl ester, ethyl nonylate, wine ether, nonanoic acid ethyl ester, natural, ethyl n-nonanoate, unii-ksh683s98j, fema no. 2447, pelargonic acid ethyl ester |
| PubChem CID | 31251 |
| ChEBI | CHEBI:87501 |
| IUPAC Name | ethyl nonanoate |
| SMILES | CCCCCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
N028925ML
|
TCI America
N028925ML |
25 mL |
Each for $48.13
|
|
|||||
|
N0289500ML
|
TCI America
N0289500ML |
500 mL |
Each for $215.62
|
|
|||||
Chemical Identifiers
| 123-29-5 | |
| 186.30 | |
| BYEVBITUADOIGY-UHFFFAOYSA-N | |
| 31251 | |
| ethyl nonanoate |
| C11H22O2 | |
| MFCD00009570 | |
| ethyl pelargonate, nonanoic acid, ethyl ester, ethyl nonylate, wine ether, nonanoic acid ethyl ester, natural, ethyl n-nonanoate, unii-ksh683s98j, fema no. 2447, pelargonic acid ethyl ester | |
| CHEBI:87501 | |
| CCCCCCCCC(=O)OCC |
Specifications
| 123-29-5 | |
| Colorless | |
| C11H22O2 | |
| 25 mL | |
| BYEVBITUADOIGY-UHFFFAOYSA-N | |
| ethyl nonanoate | |
| 31251 | |
| 186.30 | |
| Liquid |
| -44°C | |
| 220°C | |
| MFCD00009570 | |
| ethyl pelargonate, nonanoic acid, ethyl ester, ethyl nonylate, wine ether, nonanoic acid ethyl ester, natural, ethyl n-nonanoate, unii-ksh683s98j, fema no. 2447, pelargonic acid ethyl ester | |
| CCCCCCCCC(=O)OCC | |
| 186.30 | |
| CHEBI:87501 | |
| ≥95.0% (GC) | |
| Ethyl Nonanoate |
Safety and Handling
EINECSNumber : (2)-0782&(2)-0798
RTECSNumber : RA6845000
TSCA : Yes