missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Nonanoate 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | -44°C |
| Color | Colorless |
| Boiling Point | 220°C |
| Quantity | 25 mL |
| Formula Weight | 186.30 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl Nonanoate |
Safety and Handling
| EINECSNumber | (2)-0782&(2)-0798 |
| RTECSNumber | RA6845000 |
| TSCA | Yes |