missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nonanoate, 97%

Catalog No. AAB24190AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB24190AP 500 mL
AAB24190AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB24190AP Supplier Thermo Scientific Chemicals Supplier No. B24190AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123-29-5 | C11H22O2 | 186.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-29-5
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009570
InChI Key BYEVBITUADOIGY-UHFFFAOYSA-N
Synonym ethyl pelargonate, nonanoic acid, ethyl ester, ethyl nonylate, wine ether, nonanoic acid ethyl ester, natural, ethyl n-nonanoate, unii-ksh683s98j, fema no. 2447, pelargonic acid ethyl ester
PubChem CID 31251
ChEBI CHEBI:87501
IUPAC Name ethyl nonanoate
SMILES CCCCCCCCC(=O)OCC

Specifications

Melting Point -44°C
Density 0.866
Boiling Point 227°C to 229°C
Flash Point 94°C (201°F)
Odor Pleasant
Refractive Index 1.422
Linear Formula CH3(CH2)7CO2CH2CH3
Quantity 500 mL
Beilstein 1759169
Merck Index 14,3838
Formula Weight 186.3
Percent Purity 97%
Chemical Name or Material Ethyl nonanoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 204-615-7
RTECSNumber RA6845000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only