missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nonafluorovalerate 98.0+%, TCI America™

Catalog No. N06895G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06895G Supplier TCI America Supplier No. N06895G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 424-36-2
Molecular Formula C7H5F9O2
Molecular Weight (g/mol) 292.101
MDL Number MFCD00077523
InChI Key JBEYNXOZKKQLOH-UHFFFAOYSA-N
Synonym Ethyl Perfluorovalerate, Nonafluorovaleric Acid Ethyl Ester, Perfluorovaleric Acid Ethyl Ester
PubChem CID 526456
IUPAC Name ethyl 2,2,3,3,4,4,5,5,5-nonafluoropentanoate
SMILES CCOC(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 3272
Formula Weight 292.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Nonafluorovalerate